Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122838.1 | NP_001116310.1 | 4 | 1324 | ||
NM_198990.4 | NP_945341.3 | 4 | 1324 | |||
XM_005250214.4 | XP_005250271.2 | 4 | 1056 | |||
XM_005250215.2 | XP_005250272.1 | 4 | 1189 | |||
XM_005250218.2 | XP_005250275.1 | 4 | 1156 | |||
XM_011515933.2 | XP_011514235.1 | 4 | 1056 | |||
XM_011515934.2 | XP_011514236.1 | 4 | 805 | |||
XM_017011854.1 | XP_016867343.1 | 4 | 1447 | |||
XM_017011855.1 | XP_016867344.1 | 4 | 1189 | |||
GenBank mRNA | AB112352.1 | - | 3 | 996 | ||
AK304644.1 | - | 4 | 1294 | |||
AY357337.1 | - | 4 | 1090 | |||
BC037350.2 | - | 1 | 221 | |||
BC057294.1 | - | 1 | 221 | |||
BC071604.1 | - | 4 | 1286 | 4375696 | ||
CR936639.1 | - | 4 | 1274 |