Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286579.1 | NP_001273508.1 | 5 | 912 | ||
NM_001286580.1 | NP_001273509.1 | 7 | 1136 | |||
NM_173562.4 | NP_775833.2 | 8 | 1410 | |||
NR_104480.1 | 6 | 1015 | ||||
NR_104481.1 | 4 | 706 | ||||
XM_005248940.4 | XP_005248997.1 | 9 | 1265 | |||
XM_006715023.2 | XP_006715086.1 | 8 | 1183 | |||
XM_011514398.2 | XP_011512700.1 | 9 | 1261 | |||
GenBank mRNA | AK295717.1 | - | 4 | 456 | ||
AK302861.1 | - | 7 | 867 | |||
AK304377.1 | - | 8 | 1131 | |||
AK316456.1 | - | 6 | 761 | |||
AL832381.1 | - | 7 | 922 | |||
AY305862.1 | - | 8 | 1410 | |||
BC023525.2 | - | 5 | 650 | 4128926 |