Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134450.1 | NP_001127922.1 | 1, 2 | 279 | ||
NM_152913.2 | NP_690877.1 | 1, 2 | 279 | |||
GenBank mRNA | AK225783.1 | - | 1, 2 | 222 | ||
AK312364.1 | - | 1, 2 | 149 | |||
AL834348.1 | - | 1, 2 | 125 | |||
AL834349.1 | - | 1, 2 | 95 | |||
AY358583.1 | - | 2, 3 | 211 | |||
BC030793.1 | - | 1, 2 | 248 | 5263130 | ||
DA144530.1 | - | 1, 2 | 279 |