Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134450.1 | NP_001127922.1 | 4 | 807 | ||
NM_001134451.1 | NP_001127923.1 | 3 | 501 | |||
NM_152913.2 | NP_690877.1 | 4 | 807 | |||
GenBank mRNA | AK124634.1 | - | 4 | 1684 | ||
AK225783.1 | - | 4 | 750 | |||
AK294658.1 | - | 4 | 623 | |||
AK295543.1 | - | 3 | 465 | |||
AK312364.1 | - | 4 | 677 | |||
AL832523.1 | - | 3 | 541 | |||
AL834348.1 | - | 4 | 653 | |||
AL834349.1 | - | 4 | 623 | |||
AY358583.1 | - | 5 | 739 | |||
BC030793.1 | - | 4 | 776 | 5263130 | ||
BC037895.1 | - | 3 | 413 | 5275566 | ||
DA154119.1 | - | 3 | 520 |