Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152754.2 | NP_689967.2 | 17 | 2260 | ||
XM_011515960.1 | XP_011514262.1 | 19 | 2570 | |||
XM_011515961.2 | XP_011514263.1 | 17 | 2095 | |||
XM_017011873.1 | XP_016867362.1 | 18 | 2759 | |||
GenBank mRNA | AY358937.1 | - | 18 | 2326 | ||
BC029590.1 | - | 10 | 1346 | 5272175 |