Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152565.1 | NP_689778.1 | 2, 3 | 379 | ||
GenBank mRNA | AK096027.1 | - | 2, 3 | 379 | ||
AY079172.1 | - | 2, 3 | 360 | |||
BC065207.1 | - | 2, 3 | 385 | 5190344 |