Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286256.1 | NP_001273185.1 | 8, 9 | 952 | ||
NM_001286257.1 | NP_001273186.1 | 6, 7 | 586 | |||
NM_145064.2 | NP_659501.1 | 9, 10 | 1019 | |||
NR_104422.1 | 5, 6 | 519 | ||||
XM_011538126.2 | XP_011536428.1 | 9, 10 | 1107 | |||
GenBank mRNA | AK057013.1 | - | 9, 10 | 1012 | ||
AK300688.1 | - | 6, 7 | 516 | |||
AK311513.1 | - | 5, 6 | 512 | |||
BC008069.2 | - | 8, 9 | 932 | 2961089 | ||
BU738507.1 | - | 3 | 646 |