Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145659.3 | NP_663634.2 | 3 | 293 | ||
XM_011545780.2 | XP_011544082.1 | 3 | 647 | |||
GenBank mRNA | AY099296.1 | - | 3 | 270 | ||
BC062422.1 | - | 3 | 296 | 4703457 |