Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 5 | 481 | ||
NM_001288565.1 | NP_001275494.1 | 4 | 984 | |||
NM_001288566.1 | NP_001275495.1 | 5 | 582 | |||
NM_001288567.1 | NP_001275496.1 | 5 | 796 | |||
NM_001288568.1 | NP_001275497.1 | 5 | 634 | |||
NM_001288569.1 | NP_001275498.1 | 5 | 495 | |||
NM_001288570.1 | NP_001275499.1 | 5 | 508 | |||
NM_001288571.1 | NP_001275500.1 | 5 | 574 | |||
NM_016456.4 | NP_057540.1 | 5 | 499 | |||
NR_109920.1 | 6 | 726 | ||||
NR_109921.1 | 4 | 953 | ||||
XM_011509385.1 | XP_011507687.1 | 5 | 558 | |||
XM_011509386.2 | XP_011507688.1 | 5 | 533 | |||
XM_011509387.2 | XP_011507689.1 | 5 | 421 | |||
GenBank mRNA | AF151020.1 | - | 6 | 471 | ||
AK054883.1 | - | 5 | 454 | |||
BM928361.1 | - | 6 | 648 | |||
BX337142.2 | - | 4 | 837 |