Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171940.1 | NP_001165411.2 | 3, 4 | 471 | ||
NM_001171941.2 | NP_001165412.1 | 3, 4 | 405 | |||
NM_153756.2 | NP_715637.2 | 3, 4 | 471 | |||
GenBank mRNA | AK092102.1 | - | 3, 4 | 404 | ||
BC062297.1 | - | 3, 4 | 394 | 6338770 | ||
BE502835.1 | - | 3, 4 | 472 | |||
BX537781.1 | - | 3, 4 | 347 | |||
DA393478.1 | - | 1, 2 | 75 |