Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167674.1 | NP_001161146.1 | 9 | 1751 | ||
NM_001167675.1 | NP_001161147.1 | 9 | 1658 | |||
NM_001256502.1 | NP_001243431.1 | 10 | 1298 | |||
NM_001256503.1 | NP_001243432.1 | 9 | 1148 | |||
NM_001256504.1 | NP_001243433.1 | 10 | 1205 | |||
NM_001256505.1 | NP_001243434.1 | 9 | 1178 | |||
NM_153184.3 | NP_694854.2 | 9 | 1131 | |||
XM_006713081.3 | XP_006713144.1 | 10 | 1641 | |||
XM_006713082.3 | XP_006713145.1 | 8 | 1624 | |||
XM_017006062.1 | XP_016861551.1 | 11 | 1683 | |||
XM_017006063.1 | XP_016861552.1 | 8 | 1211 | |||
GenBank mRNA | AB646739.1 | - | 9 | 1658 | ||
AB646740.1 | - | 9 | 1178 | |||
AB646741.1 | - | 10 | 1298 | |||
AB646742.1 | - | 10 | 1205 | |||
AB646743.1 | - | 9 | 1148 | |||
AF538973.1 | - | 9 | 1131 | |||
AL833049.1 | - | 6 | 695 | |||
AL834270.1 | - | 9 | 1721 | |||
BC105999.1 | - | 9 | 1658 | 5259422 | ||
BC112136.1 | - | 9 | 1187 | 8327604 | ||
BC112138.1 | - | 9 | 1199 | 8327606 |