Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278505.1 | NP_001265434.1 | 3, 4 | 310 | ||
NM_001322249.1 | NP_001309178.1 | 4, 5 | 537 | |||
NM_174931.3 | NP_777591.3 | 5, 6 | 579 | |||
XM_017003748.1 | XP_016859237.1 | 5, 6 | 480 | |||
GenBank mRNA | AK095667.1 | - | 3, 4 | 243 | ||
AK289504.1 | - | 3, 4 | 225 | |||
AK294697.1 | - | 5, 6 | 493 | |||
BC071798.1 | - | 3, 4 | 242 | |||
BM285380.1 | - | 5, 6 | 579 |