Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005463.2 | NP_001005463.1 | 15, 16 | 1673 | ||
XM_005252668.3 | XP_005252725.1 | 15, 16 | 2255 | |||
XM_005252669.3 | XP_005252726.1 | 15, 16 | 2228 | |||
XM_006717743.3 | XP_006717806.1 | 15, 16 | 2147 | |||
XM_017016027.1 | XP_016871516.1 | 14, 15 | 1958 | |||
GenBank mRNA | AL832828.1 | - | 15, 16 | 1600 | ||
BC126130.1 | - | 15, 16 | 1664 | 8991846 | ||
BC130479.1 | - | 15, 16 | 1664 | 40146510 |