Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 3 | 510 | ||
NM_153251.3 | NP_694983.2 | 4 | 536 | |||
NR_104486.1 | 4 | 537 | ||||
NR_104487.1 | 4 | 537 | ||||
XM_006719795.1 | XP_006719858.1 | 4 | 476 | |||
XM_006719796.1 | XP_006719859.1 | 4 | 476 | |||
XM_006719797.1 | XP_006719860.1 | 4 | 476 | |||
XM_017020525.1 | XP_016876014.1 | 2 | 169 | |||
XM_017020526.1 | XP_016876015.1 | 5 | 242 | |||
XM_017020527.1 | XP_016876016.1 | 4 | 452 | |||
GenBank mRNA | AK295804.1 | - | 4 | 442 | ||
AK299354.1 | - | 3 | 448 | |||
BC034944.1 | - | 4 | 542 | |||
BC050367.1 | - | 4 | 451 | 5591379 | ||
BC067898.1 | - | 4 | 536 | |||
BC110517.1 | - | 4 | 415 | 40032074 | ||
BG720734.1 | - | 4 | 543 |