Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 6, 7 | 773 | ||
NM_153251.3 | NP_694983.2 | 7, 8 | 799 | |||
NR_104486.1 | 8, 9 | 918 | ||||
NR_104487.1 | 7, 8 | 800 | ||||
XM_006719795.1 | XP_006719858.1 | 7, 8 | 739 | |||
XM_006719796.1 | XP_006719859.1 | 7, 8 | 739 | |||
XM_006719797.1 | XP_006719860.1 | 7, 8 | 739 | |||
XM_017020525.1 | XP_016876014.1 | 5, 6 | 432 | |||
XM_017020526.1 | XP_016876015.1 | 8, 9 | 505 | |||
XM_017020527.1 | XP_016876016.1 | 7, 8 | 715 | |||
GenBank mRNA | AK299354.1 | - | 6, 7 | 711 | ||
BC034944.1 | - | 8, 9 | 923 | |||
BC050367.1 | - | 7, 8 | 714 | 5591379 | ||
BC067898.1 | - | 8, 9 | 917 | |||
BC110517.1 | - | 7, 8 | 678 | 40032074 |