Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGAPA1 SIRNA Assays ›
Gene Symbol:
RALGAPA1
RALGAPA1P1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
Chromosome Location:
Chr.9: 105519824 - 105527319 on Build GRCh38
UniGene ID:
Hs.113150
Hs.113150
Species:
Human
siRNA ID
HSS137789
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGAPA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283043.1
NP_001269972.1
12
1861
NM_001283044.1
NP_001269973.1
12
1861
NM_014990.1
NP_055805.1
12
1861
NM_194301.2
NP_919277.2
12
1861
XM_005267491.3
XP_005267548.1
12
1939
XM_005267492.3
XP_005267549.1
12
1939
XM_005267493.3
XP_005267550.1
12
1939
XM_006720098.2
XP_006720161.1
12
1939
XM_006720099.2
XP_006720162.1
12
1939
XM_006720100.3
XP_006720163.1
12
1939
XM_006720101.2
XP_006720164.1
12
1939
XM_006720102.3
XP_006720165.1
12
1645
XM_006720103.2
XP_006720166.1
12
1939
XM_006720104.3
XP_006720167.1
12
1939
XM_011536615.2
XP_011534917.1
12
1939
XM_011536616.2
XP_011534918.1
12
1939
XM_017021141.1
XP_016876630.1
12
1939
XM_017021142.1
XP_016876631.1
12
1939
XM_017021143.1
XP_016876632.1
12
1939
XM_017021144.1
XP_016876633.1
12
1939
XM_017021145.1
XP_016876634.1
12
1939
GenBank mRNA
AB020691.1
-
7
989
AB511280.1
-
12
1470
AK022988.1
-
7
952
AY596970.1
-
12
1861
AY596971.1
-
12
1861
BC016305.1
-
7
1005
BC150596.1
-
12
1569
9056966
Gene Symbol: RALGAPA1P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_104269.1
1
1900
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline