Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322278.1 | NP_001309207.1 | 6 | 722 | ||
NM_001322279.1 | NP_001309208.1 | 6 | 686 | |||
NM_199227.2 | NP_954697.1 | 6 | 636 | |||
NR_136273.1 | 5 | 573 | ||||
NR_136276.1 | 5 | 487 | ||||
XM_017003750.1 | XP_016859239.1 | 6 | 635 | |||
GenBank mRNA | AY374142.1 | - | 6 | 636 | ||
BC113644.1 | - | 6 | 581 | 8322696 | ||
BC143913.1 | - | 6 | 667 | |||
DQ005576.1 | - | 6 | 950 |