Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 24 | 3045 | ||
XM_005262113.3 | XP_005262170.1 | 23 | 2897 | |||
XM_017029384.1 | XP_016884873.1 | 20 | 2575 | |||
GenBank mRNA | AK095887.1 | - | 7 | 918 | ||
AY497046.1 | - | 24 | 3045 | |||
AY497047.1 | - | 21 | 2977 | |||
AY497048.1 | - | 24 | 3068 | |||
AY497049.1 | - | 23 | 3038 | |||
AY497050.1 | - | 24 | 3018 | |||
AY497051.1 | - | 21 | 2605 | |||
AY497052.1 | - | 26 | 3204 | |||
AY497053.1 | - | 22 | 2793 | |||
AY497054.1 | - | 21 | 2694 | |||
AY497055.1 | - | 24 | 2979 | |||
AY497056.1 | - | 21 | 2950 | |||
AY497057.1 | - | 20 | 2849 | |||
AY497058.1 | - | 20 | 2688 | |||
AY497059.1 | - | 22 | 3059 | |||
AY497060.1 | - | 18 | 2626 |