Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182553.2 | NP_872359.1 | 5 | 666 | ||
NR_073078.1 | 5 | 692 | ||||
NR_073079.1 | 5 | 662 | ||||
GenBank mRNA | AY726585.1 | - | 5 | 437 | ||
BC047953.1 | - | 5 | 642 | |||
BM547951.1 | - | 666 | ||||
DR760677.1 | - | 5 | 638 |