Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 3 | 361 | ||
NM_152770.2 | NP_689983.2 | 3 | 361 | |||
XM_011531816.2 | XP_011530118.2 | 4 | 527 | |||
XM_017007972.1 | XP_016863461.1 | 4 | 513 | |||
XM_017007973.1 | XP_016863462.1 | 4 | 516 | |||
XM_017007974.1 | XP_016863463.1 | 4 | 521 | |||
XM_017007975.1 | XP_016863464.1 | 2 | 501 | |||
XM_017007976.1 | XP_016863465.1 | 4 | 536 | |||
XM_017007977.1 | XP_016863466.1 | 4 | 527 | |||
XM_017007978.1 | XP_016863467.1 | 2 | 378 | |||
GenBank mRNA | AI650567.1 | - | 4 | 521 | ||
AK128642.1 | - | 3 | 313 | |||
AY349359.1 | - | 3 | 312 | |||
BC034296.1 | - | 3 | 361 | 5165558 |