Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153259.3 | NP_694991.2 | 2 | 347 | ||
XM_005270719.3 | XP_005270776.1 | 2 | 124 | |||
XM_006710552.2 | XP_006710615.1 | 3 | 437 | |||
XM_011541187.2 | XP_011539489.1 | 2 | 181 | |||
XM_011541188.2 | XP_011539490.1 | 2 | 464 | |||
XM_017000923.1 | XP_016856412.1 | 2 | 338 | |||
GenBank mRNA | AK094010.1 | - | 2 | 338 | ||
AK309858.1 | - | 2 | 338 | |||
AY083533.1 | - | 1 | 13 | |||
BC104891.1 | - | 2 | 165 | 8143894 | ||
BC104893.1 | - | 2 | 165 | 8143896 | ||
DR007194.1 | - | 2 | 363 |