Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNEP1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281789.1 | NP_001268718.1 | 4, 5 | 381 | ||
NM_153261.5 | NP_694993.2 | 5, 6 | 432 | |||
NR_104042.1 | 4, 5 | 358 | ||||
NR_104043.1 | 3, 4 | 307 | ||||
GenBank mRNA | AK095420.1 | - | 4, 5 | 354 | ||
AK301610.1 | - | 5, 6 | 397 | |||
AK307132.1 | - | 4, 5 | 428 | |||
AK307309.1 | - | 3, 4 | 249 | |||
BC022550.1 | - | 3, 4 | 258 | |||
BC036683.2 | - | 5, 6 | 361 | 5266084 | ||
BP259769.1 | - | 4, 5 | 330 | |||
CR933600.1 | - | 5, 6 | 377 | |||
FJ516381.1 | - | 4, 5 | 309 | |||
HY011229.1 | - | 4, 5 | 382 |