Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 4 | 417 | ||
NM_001146262.2 | NP_001139734.1 | 3 | 296 | |||
NM_001146264.2 | NP_001139736.1 | 4 | 417 | |||
NM_001256006.1 | NP_001242935.1 | 4 | 501 | |||
NM_153262.3 | NP_694994.2 | 3 | 296 | |||
NR_027459.2 | 4 | 501 | ||||
XM_006711262.3 | XP_006711325.1 | 1 | 59 | |||
XM_017000931.1 | XP_016856420.1 | 1 | 59 | |||
XM_017000932.1 | XP_016856421.1 | 4 | 491 | |||
XM_017000933.1 | XP_016856422.1 | 4 | 354 | |||
XM_017000934.1 | XP_016856423.1 | 1 | 150 | |||
XM_017000935.1 | XP_016856424.1 | 3 | 305 | |||
GenBank mRNA | AB102948.1 | - | 2 | 205 | ||
AJ617623.1 | - | 3 | 239 | |||
AJ617624.1 | - | 3 | 382 | |||
AJ617625.1 | - | 4 | 415 | |||
AK091517.1 | - | 3 | 219 | |||
BC130323.1 | - | 4 | 350 | 40146354 | ||
BC144154.1 | - | 4 | 350 | |||
BC144155.1 | - | 3 | 229 | |||
BC144157.1 | - | 4 | 350 |