Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 5, 6 | 649 | ||
NM_001146262.2 | NP_001139734.1 | 4, 5 | 528 | |||
NM_001146264.2 | NP_001139736.1 | 5, 6 | 649 | |||
NM_001256006.1 | NP_001242935.1 | 5, 6 | 733 | |||
NM_153262.3 | NP_694994.2 | 4, 5 | 528 | |||
NR_027459.2 | 5, 6 | 733 | ||||
XM_006711262.3 | XP_006711325.1 | 3, 4 | 1707 | |||
XM_017000931.1 | XP_016856420.1 | 3, 4 | 1707 | |||
XM_017000932.1 | XP_016856421.1 | 5, 6 | 723 | |||
XM_017000933.1 | XP_016856422.1 | 5, 6 | 586 | |||
XM_017000934.1 | XP_016856423.1 | 2, 3 | 382 | |||
GenBank mRNA | AB102948.1 | - | 3, 4 | 437 | ||
AJ617623.1 | - | 4, 5 | 471 | |||
AJ617624.1 | - | 4, 5 | 614 | |||
AJ617625.1 | - | 5, 6 | 647 | |||
AJ617626.1 | - | 1, 2 | 204 | |||
AK091517.1 | - | 4, 5 | 451 | |||
BC130323.1 | - | 5, 6 | 582 | 40146354 | ||
BC144154.1 | - | 5, 6 | 582 | |||
BC144155.1 | - | 4, 5 | 461 | |||
BC144157.1 | - | 5, 6 | 582 |