Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 7 | 1177 | ||
NM_001146262.2 | NP_001139734.1 | 6 | 1056 | |||
NM_001146264.2 | NP_001139736.1 | 7 | 1177 | |||
NM_001256006.1 | NP_001242935.1 | 7 | 1261 | |||
NM_153262.3 | NP_694994.2 | 6 | 1056 | |||
XM_006711262.3 | XP_006711325.1 | 5 | 2235 | |||
XM_017000931.1 | XP_016856420.1 | 5 | 2235 | |||
XM_017000932.1 | XP_016856421.1 | 7 | 1251 | |||
XM_017000933.1 | XP_016856422.1 | 7 | 1114 | |||
XM_017000934.1 | XP_016856423.1 | 4 | 910 | |||
XM_017000935.1 | XP_016856424.1 | 5 | 793 | |||
GenBank mRNA | AB102948.1 | - | 5 | 965 | ||
AJ617623.1 | - | 6 | 999 | |||
AJ617624.1 | - | 6 | 1142 | |||
AJ617626.1 | - | 3 | 732 | |||
AK091517.1 | - | 6 | 979 | |||
BC130323.1 | - | 7 | 1110 | 40146354 | ||
BC144154.1 | - | 7 | 1110 | |||
BC144155.1 | - | 6 | 989 | |||
BC144157.1 | - | 7 | 1110 |