Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300793.1 | NP_001287722.1 | 18 | 2418 | ||
NM_001300794.1 | NP_001287723.1 | 18 | 2415 | |||
NM_153265.2 | NP_694997.2 | 18 | 2415 | |||
XM_005273878.3 | XP_005273935.1 | 18 | 2673 | |||
XM_006718489.3 | XP_006718552.1 | 18 | 2790 | |||
XM_006718490.3 | XP_006718553.1 | 18 | 2790 | |||
XM_006718491.3 | XP_006718554.1 | 18 | 2787 | |||
XM_011544896.2 | XP_011543198.1 | 18 | 2787 | |||
XM_011544897.2 | XP_011543199.1 | 17 | 2634 | |||
XM_017017480.1 | XP_016872969.1 | 18 | 2161 | |||
XM_017017481.1 | XP_016872970.1 | 17 | 2047 | |||
XM_017017482.1 | XP_016872971.1 | 18 | 2161 | |||
XM_017017483.1 | XP_016872972.1 | 18 | 2218 | |||
GenBank mRNA | AK093146.1 | - | 17 | 2090 | ||
AK128679.1 | - | 17 | 2832 | |||
AK296901.1 | - | 18 | 2180 | |||
AL833388.1 | - | 15 | 1670 | |||
BC075824.1 | - | 17 | 2021 | |||
BC108280.1 | - | 18 | 2199 | 3915493 |