Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286408.1 | NP_001273337.1 | 4 | 579 | ||
NM_198081.4 | NP_932347.2 | 5 | 794 | |||
XM_011535703.2 | XP_011534005.1 | 5 | 692 | |||
XM_011535704.2 | XP_011534006.1 | 5 | 570 | |||
XM_011535706.1 | XP_011534008.1 | 4 | 536 | |||
XM_011535707.2 | XP_011534009.1 | 5 | 813 | |||
XM_011535710.2 | XP_011534012.1 | 2 | 214 | |||
XM_017010677.1 | XP_016866166.1 | 5 | 861 | |||
XM_017010678.1 | XP_016866167.1 | 5 | 774 | |||
XM_017010679.1 | XP_016866168.1 | 5 | 672 | |||
XM_017010680.1 | XP_016866169.1 | 5 | 740 | |||
XM_017010681.1 | XP_016866170.1 | 5 | 650 | |||
XM_017010682.1 | XP_016866171.1 | 5 | 864 | |||
XM_017010683.1 | XP_016866172.1 | 4 | 533 | |||
XM_017010684.1 | XP_016866173.1 | 5 | 866 | |||
XM_017010685.1 | XP_016866174.1 | 4 | 660 | |||
XM_017010686.1 | XP_016866175.1 | 4 | 645 | |||
XM_017010687.1 | XP_016866176.1 | 5 | 862 | |||
XM_017010688.1 | XP_016866177.1 | 5 | 870 | |||
GenBank mRNA | AK074125.1 | - | 3 | 2119 | ||
AK093571.1 | - | 4 | 579 | |||
AK303562.1 | - | 5 | 781 | |||
BC033286.1 | - | 5 | 561 | 4509724 | ||
CR975586.1 | - | 4 | 424 |