Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7D2 SIRNA Assays ›
Gene Symbol:
MAP7D2
Gene Name:
MAP7 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 20006713 - 20116996 on Build GRCh38
UniGene ID:
Hs.127951
Species:
Human
siRNA ID
HSS138016
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7D2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168465.1
NP_001161937.1
15
2119
NM_001168466.1
NP_001161938.1
13
1861
NM_001168467.1
NP_001161939.1
14
1833
NM_152780.3
NP_689993.2
14
1996
XM_005274478.2
XP_005274535.1
15
1982
XM_005274479.2
XP_005274536.1
15
1949
XM_005274480.3
XP_005274537.1
14
1883
XM_006724481.2
XP_006724544.1
16
2129
XM_006724482.2
XP_006724545.1
15
2006
XM_011545483.1
XP_011543785.1
16
2096
XM_011545484.1
XP_011543786.1
16
2079
XM_011545485.1
XP_011543787.1
16
2014
XM_017029393.1
XP_016884882.1
15
1973
XM_017029394.1
XP_016884883.1
15
1956
XM_017029395.1
XP_016884884.1
15
1932
XM_017029396.1
XP_016884885.1
15
1891
XM_017029397.1
XP_016884886.1
15
1867
GenBank mRNA
AK027409.1
-
14
1898
AK127056.1
-
12
1535
AK294788.1
-
14
1827
AK296319.1
-
13
1772
AK303019.1
-
9
1107
AK316117.1
-
13
1769
BC037165.2
-
12
1663
BC089400.1
-
15
2040
3063898
BC136379.1
-
14
1913
9020366
KP849473.1
-
15
1954
KP849474.1
-
15
1921
KP979782.1
-
15
1988
KP979783.1
-
15
1955
KP979784.1
-
15
2012
KP979785.1
-
15
1979
KP979786.1
-
15
2012
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline