Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174071.2 | NP_001167542.1 | 3 | 459 | ||
NM_001174072.2 | NP_001167543.1 | 3 | 459 | |||
NM_178276.6 | NP_840060.1 | 3 | 459 | |||
NR_126060.1 | 3 | 459 | ||||
NR_126061.1 | 3 | 459 | ||||
XM_011543304.1 | XP_011541606.1 | 3 | 424 | |||
XM_017009323.1 | XP_016864812.1 | 3 | 405 | |||
XM_017009324.1 | XP_016864813.1 | 3 | 424 | |||
XM_017009325.1 | XP_016864814.1 | 2 | 272 | |||
XM_017009326.1 | XP_016864815.1 | 2 | 272 | |||
GenBank mRNA | AF498273.1 | - | 3 | 429 | ||
AK297467.1 | - | 3 | 456 | |||
BC101280.2 | - | 3 | 391 | 40024444 | ||
BC101281.3 | - | 3 | 396 | 40024445 | ||
BC101283.2 | - | 3 | 396 | 40024448 | ||
DC354040.1 | - | 3 | 459 |