Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152784.3 | NP_689997.3 | 21 | 2088 | ||
XM_011527885.2 | XP_011526187.1 | 22 | 2578 | |||
XM_011527886.2 | XP_011526188.1 | 22 | 2524 | |||
XM_011527887.2 | XP_011526189.1 | 21 | 2449 | |||
XM_011527891.1 | XP_011526193.1 | 13 | 1374 | |||
XM_011527892.1 | XP_011526194.1 | 11 | 1095 | |||
XM_017026564.1 | XP_016882053.1 | 21 | 2494 | |||
XM_017026565.1 | XP_016882054.1 | 21 | 2231 | |||
XM_017026566.1 | XP_016882055.1 | 18 | 2164 | |||
GenBank mRNA | BC031233.1 | - | 10 | 1038 | ||
BC043005.1 | - | 21 | 2074 | 5297674 |