Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114759.2 | NP_001108231.1 | 6 | 1521 | ||
NM_001307925.1 | NP_001294854.1 | 7 | 1578 | |||
NM_173574.3 | NP_775845.2 | 6 | 1518 | |||
XM_005245828.3 | XP_005245885.1 | 7 | 1695 | |||
XM_005245830.3 | XP_005245887.1 | 7 | 1576 | |||
XM_005245832.3 | XP_005245889.1 | 7 | 1601 | |||
XM_006710555.3 | XP_006710618.1 | 8 | 1779 | |||
XM_011541198.2 | XP_011539500.1 | 7 | 1598 | |||
XM_017000954.1 | XP_016856443.1 | 7 | 1698 | |||
XM_017000955.1 | XP_016856444.1 | 7 | 1576 | |||
XM_017000956.1 | XP_016856445.1 | 7 | 1573 | |||
XM_017000957.1 | XP_016856446.1 | 6 | 1638 | |||
XM_017000958.1 | XP_016856447.1 | 7 | 1678 | |||
XM_017000959.1 | XP_016856448.1 | 6 | 1618 | |||
GenBank mRNA | BC028731.2 | - | 6 | 1462 | 4829037 | |
BC029505.1 | - | 7 | 1559 | 5273528 |