Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 4 | 2254 | ||
NM_018136.4 | NP_060606.3 | 4 | 2254 | |||
GenBank mRNA | AF509326.1 | - | 4 | 1997 | ||
AK125107.1 | - | 4 | 2254 | |||
AK226178.1 | - | 4 | 2187 | |||
AY099891.1 | - | 2 | 779 | |||
AY099893.1 | - | 2 | 779 | |||
AY101201.1 | - | 3 | 1664 | |||
AY367065.1 | - | 4 | 1997 | |||
AY971956.1 | - | 4 | 1997 |