Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320289.1 | NP_001307218.1 | 12 | 1732 | ||
NM_001320290.1 | NP_001307219.1 | 12 | 1469 | |||
NM_178033.1 | NP_828847.1 | 12 | 1766 | |||
GenBank mRNA | AK091806.1 | - | 12 | 1470 | ||
AK098065.1 | - | 12 | 1759 | |||
AK131355.1 | - | 12 | 1732 | |||
AM040940.1 | - | 12 | 1603 | |||
AY358537.1 | - | 12 | 1749 | |||
BC028102.1 | - | 12 | 1766 | 5243005 |