Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077241.2 | NP_001070709.2 | 4 | 347 | ||
NM_001278250.2 | NP_001265179.2 | 3 | 558 | |||
NM_001278251.2 | NP_001265180.2 | 8 | 1204 | |||
NM_182556.3 | NP_872362.3 | 5 | 402 | |||
XM_006718509.3 | XP_006718572.1 | 4 | 1012 | |||
XM_006718510.3 | XP_006718573.1 | 6 | 540 | |||
XM_011544943.2 | XP_011543245.1 | 5 | 1111 | |||
XM_011544944.2 | XP_011543246.1 | 4 | 441 | |||
XM_011544947.2 | XP_011543249.1 | 4 | 630 | |||
XM_011544949.2 | XP_011543251.1 | 6 | 459 | |||
XM_017017562.1 | XP_016873051.1 | 4 | 630 | |||
XM_017017563.1 | XP_016873052.1 | 5 | 1877 | |||
XM_017017564.1 | XP_016873053.1 | 5 | 1876 | |||
XM_017017565.1 | XP_016873054.1 | 5 | 1141 | |||
XM_017017566.1 | XP_016873055.1 | 5 | 496 | |||
XM_017017567.1 | XP_016873056.1 | 6 | 502 | |||
XM_017017568.1 | XP_016873057.1 | 5 | 325 | |||
XM_017017569.1 | XP_016873058.1 | 7 | 1204 | |||
GenBank mRNA | AK090434.1 | - | 1 | 159 | ||
AK091190.1 | - | 8 | 1204 | |||
BC036869.1 | - | 4 | 310 | 5228549 | ||
BX648573.1 | - | 2 | 2934 | |||
DA872876.1 | - | 5 | 399 | |||
DB049668.1 | - | 5 | 403 |