Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182557.2 | NP_872363.1 | 6 | 3714 | ||
XM_005271511.2 | XP_005271568.1 | 8 | 3714 | |||
XM_005271512.2 | XP_005271569.1 | 7 | 3164 | |||
XM_005271513.2 | XP_005271570.1 | 6 | 3163 | |||
XM_006718815.2 | XP_006718878.1 | 7 | 3212 | |||
XM_011542756.2 | XP_011541058.1 | 6 | 3670 | |||
XM_017017572.1 | XP_016873061.1 | 8 | 3963 | |||
XM_017017573.1 | XP_016873062.1 | 7 | 4315 | |||
XM_017017574.1 | XP_016873063.1 | 7 | 3862 | |||
GenBank mRNA | AB094091.1 | - | 6 | 3714 | ||
AK122650.1 | - | 1 | 158 | |||
AK122882.1 | - | 1 | 1354 | |||
AY296059.1 | - | 6 | 2734 | |||
BC033257.1 | - | 1 | 633 |