Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198474.3 | NP_940876.2 | 3 | 1159 | ||
XM_005252878.4 | XP_005252935.1 | 4 | 936 | |||
GenBank mRNA | AK297778.1 | - | 3 | 657 | ||
AK298137.1 | - | 4 | 754 | |||
AY358591.1 | - | 3 | 761 | |||
BC035895.1 | - | 3 | 986 | 5229159 | ||
BC092488.1 | - | 3 | 1004 | 30531479 | ||
BX647908.1 | - | 3 | 1047 |