Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004304.3 | NP_001004304.1 | 5 | 721 | ||
XM_006719344.2 | XP_006719407.1 | 5 | 696 | |||
XM_011538194.2 | XP_011536496.1 | 5 | 921 | |||
XM_017019181.1 | XP_016874670.1 | 5 | 955 | |||
GenBank mRNA | AK091946.1 | - | 2 | 104 | ||
AK292744.1 | - | 5 | 772 | |||
BC053557.1 | - | 5 | 724 | 6162269 |