Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135195.1 | NP_001128667.1 | 3, 4 | 688 | ||
NM_173596.2 | NP_775867.2 | 5, 6 | 717 | |||
XM_005268803.1 | XP_005268860.1 | 3, 4 | 605 | |||
XM_011538198.1 | XP_011536500.1 | 3, 4 | 670 | |||
XM_011538199.1 | XP_011536501.1 | 5, 6 | 724 | |||
XM_011538200.1 | XP_011536502.1 | 4, 5 | 674 | |||
XM_011538201.1 | XP_011536503.1 | 4, 5 | 648 | |||
XM_017019185.1 | XP_016874674.1 | 3, 4 | 648 | |||
XM_017019186.1 | XP_016874675.1 | 5, 6 | 835 | |||
XM_017019187.1 | XP_016874676.1 | 3, 4 | 584 | |||
GenBank mRNA | AK172768.1 | - | 3, 4 | 651 | ||
AK298331.1 | - | 3, 4 | 329 | |||
AK313188.1 | - | 5, 6 | 680 | |||
BC027884.1 | - | 3, 4 | 562 | 5221329 |