Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182559.2 | NP_872365.1 | 5 | 929 | ||
GenBank mRNA | BC048112.1 | - | 5 | 914 | 4830541 |