Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135919.1 | NP_001129391.1 | 4, 5 | 1696 | ||
NM_181785.3 | NP_861450.1 | 4, 5 | 1696 | |||
XM_005266361.2 | XP_005266418.1 | 4, 5 | 1374 | |||
GenBank mRNA | AF315594.1 | - | 4, 5 | 1280 | ||
AK026720.1 | - | 1 | 475 | |||
AK093932.1 | - | 3, 4 | 2124 | |||
AK098426.1 | - | 2, 3 | 726 | |||
AK124604.1 | - | 2, 3 | 2389 | |||
BC036662.1 | - | 4, 5 | 1658 | 4827143 | ||
BC068556.1 | - | 4, 5 | 1524 | 4820792 | ||
BX537565.1 | - | 4, 5 | 1507 | |||
CB851220.1 | - | 2, 3 | 223 |