Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213601.1 | NP_998766.1 | 4 | 370 | ||
XM_017021223.1 | XP_016876712.1 | 4 | 716 | |||
XM_017021224.1 | XP_016876713.1 | 3 | 269 | |||
XM_017021225.1 | XP_016876714.1 | 4 | 2831 | |||
GenBank mRNA | AK095650.1 | - | 4 | 414 | ||
AY598329.1 | - | 4 | 370 | |||
BC101510.1 | - | 4 | 426 | 8069016 |