Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173611.3 | NP_775882.2 | 3, 4 | 375 | ||
GenBank mRNA | AK095745.1 | - | 3, 4 | 349 | ||
BC035600.1 | - | 2, 3 | 233 | |||
BC093898.1 | - | 3, 4 | 345 | 7939743 | ||
BP872319.1 | - | 3, 4 | 375 | |||
BP872410.1 | - | 3, 4 | 375 |