Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130918.1 | NP_001124390.1 | 10 | 1410 | ||
NM_173623.3 | NP_775894.2 | 3 | 498 | |||
XM_017024491.1 | XP_016879980.1 | 10 | 1553 | |||
XM_017024492.1 | XP_016879981.1 | 9 | 1666 | |||
XM_017024493.1 | XP_016879982.1 | 11 | 1620 | |||
XM_017024494.1 | XP_016879983.1 | 10 | 1555 | |||
XM_017024495.1 | XP_016879984.1 | 9 | 2283 | |||
XM_017024496.1 | XP_016879985.1 | 5 | 693 | |||
XM_017024497.1 | XP_016879986.1 | 10 | 1975 | |||
GenBank mRNA | AK093127.1 | - | 2 | 362 | ||
AK097392.1 | - | 10 | 1322 | |||
AK302952.1 | - | 10 | 1356 | |||
AK310321.1 | - | 3 | 508 | |||
AL834151.1 | - | 10 | 1976 | |||
BC033563.1 | - | 3 | 499 | 4825815 | ||
BC041368.2 | - | 10 | 2406 |