Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213602.2 | NP_998767.1 | 4 | 847 | ||
GenBank mRNA | AK092746.1 | - | 1 | 398 | ||
AK095432.1 | - | 5 | 551 | |||
AK172835.1 | - | 4 | 847 | |||
AK290400.1 | - | 4 | 701 | |||
AK290402.1 | - | 4 | 711 | |||
AK301189.1 | - | 4 | 728 | |||
AY971516.1 | - | 4 | 836 |