Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 564 | ||
NM_175907.5 | NP_787103.1 | 2 | 742 | |||
GenBank mRNA | AK292880.1 | - | 2 | 497 | ||
AK302801.1 | - | 2 | 294 | |||
BC018081.1 | - | 2 | 720 | |||
BC033780.1 | - | 2 | 628 | 4510603 | ||
BC078661.1 | - | 2 | 555 | 5575156 | ||
BX361234.2 | - | 2 | 744 | |||
BX647730.1 | - | 2 | 565 |