Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199324.1 | NP_001186253.1 | 7 | 726 | ||
NM_001321317.1 | NP_001308246.1 | 4 | 451 | |||
NM_001321318.1 | NP_001308247.1 | 6 | 522 | |||
NM_001321319.1 | NP_001308248.1 | 6 | 688 | |||
NM_001321320.1 | NP_001308249.1 | 5 | 655 | |||
NM_001321321.1 | NP_001308250.1 | 6 | 681 | |||
NM_001321322.1 | NP_001308251.1 | 6 | 676 | |||
NM_001321323.1 | NP_001308252.1 | 5 | 484 | |||
NM_198480.3 | NP_940882.3 | 6 | 693 | |||
XM_011526824.1 | XP_011525126.1 | 8 | 1004 | |||
XM_011526825.1 | XP_011525127.1 | 8 | 1130 | |||
XM_011526826.1 | XP_011525128.1 | 7 | 970 | |||
XM_011526829.1 | XP_011525131.1 | 6 | 739 | |||
XM_011526830.1 | XP_011525132.1 | 5 | 542 | |||
XM_017026648.1 | XP_016882137.1 | 7 | 971 | |||
XM_017026649.1 | XP_016882138.1 | 7 | 1097 | |||
XM_017026650.1 | XP_016882139.1 | 7 | 804 | |||
XM_017026651.1 | XP_016882140.1 | 8 | 1217 | |||
GenBank mRNA | AK131494.1 | - | 6 | 668 | ||
BC053884.1 | - | 6 | 761 | |||
BC105100.1 | - | 4 | 414 | 8144103 | ||
BC105102.1 | - | 4 | 414 | 8144105 | ||
BC143429.1 | - | 5 | 447 | 9051937 | ||
BX647556.1 | - | 6 | 654 |