Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302813.1 | NP_001289742.1 | 3 | 320 | ||
NM_173644.2 | NP_775915.1 | 4 | 400 | |||
XM_017027828.1 | XP_016883317.1 | 3 | 9125 | |||
XM_017027829.1 | XP_016883318.1 | 4 | 6787 | |||
XM_017027830.1 | XP_016883319.1 | 3 | 6652 | |||
XM_017027831.1 | XP_016883320.1 | 4 | 8471 | |||
XM_017027832.1 | XP_016883321.1 | 4 | 524 | |||
GenBank mRNA | AK091179.1 | - | 4 | 400 | ||
AY358539.1 | - | 1 | 2109 | |||
BC101349.1 | - | 3 | 266 | 40025393 | ||
BC101351.2 | - | 3 | 266 | 40025395 | ||
CR744567.1 | - | 3 | 318 |