Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161368.1 | NP_001154840.1 | 6 | 544 | ||
NM_174977.3 | NP_777637.1 | 6 | 544 | |||
XM_006724235.2 | XP_006724298.1 | 6 | 560 | |||
XM_011530164.2 | XP_011528466.1 | 6 | 724 | |||
XM_017028787.1 | XP_016884276.1 | 5 | 673 | |||
XM_017028788.1 | XP_016884277.1 | 5 | 674 | |||
XM_017028789.1 | XP_016884278.1 | 5 | 346 | |||
XM_017028790.1 | XP_016884279.1 | 5 | 681 | |||
XM_017028791.1 | XP_016884280.1 | 5 | 373 | |||
GenBank mRNA | AK093418.1 | - | 6 | 561 | ||
AK093448.1 | - | 6 | 528 | |||
AY158085.1 | - | 6 | 444 | |||
BC136358.1 | - | 6 | 516 | 9020345 | ||
BC136359.1 | - | 6 | 516 | 9020346 | ||
BC139912.1 | - | 6 | 483 | 40147686 | ||
CR456346.1 | - | 4 | 303 |