Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173651.3 | NP_775922.3 | 7 | 787 | ||
GenBank mRNA | AK092099.1 | - | 3 | 199 | ||
BC121088.1 | - | 3 | 203 | |||
BC121089.1 | - | 3 | 203 | |||
BC128052.1 | - | 3 | 203 | |||
BC148281.2 | - | 3 | 203 | |||
CR936597.1 | - | 2 | 143 |