Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173653.3 | NP_775924.1 | 5 | 840 | ||
XM_011512703.2 | XP_011511005.1 | 1 | 52 | |||
XM_011512704.2 | XP_011511006.1 | 5 | 813 | |||
XM_017006202.1 | XP_016861691.1 | 5 | 813 | |||
XM_017006203.1 | XP_016861692.1 | 4 | 598 | |||
GenBank mRNA | AB089794.1 | - | 5 | 631 | ||
AK301816.1 | - | 4 | 566 | |||
AK315107.1 | - | 5 | 767 | |||
AL832304.1 | - | 5 | 705 | |||
AY254100.1 | - | 5 | 755 | |||
BC035779.1 | - | 5 | 756 | 5590356 |